2-Mercaptopyrimidine
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2-Thiopyrimidine
- CAS Number:1450-85-7
- Formula:C₄H₄N₂S
- MDL Number:MFCD00006073
- Molecular Weight:112.16 g/mol
- Melting Pt:230 °C (dec.)
- Storage Temperature:Ambient
- EINECS:215-917-3
6 Options Available Below
Product Details & Documents
Synonyms: 2-Thiopyrimidine
- CAS Number:1450-85-7
- Formula:C₄H₄N₂S
- MDL Number:MFCD00006073
- Molecular Weight:112.16 g/mol
- Melting Pt:230 °C (dec.)
- Storage Temperature:Ambient
- EINECS:215-917-3
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



