Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,6-dichloroisonicotinate
Methyl-2,6-dichloroisonicotinate
Methyl-2,6-dichloroisonicotinate
Methyl-2,6-dichloroisonicotinate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-2,6-dichloroisonicotinate
Supplier:
Apollo Scientific
About this item
CAS Number:
42521-09-5
:
Formula:
C₇H₅Cl₂NO₂
MDL Number:
MFCD00044409
:
206.03 g/mol
Melting Pt:
80…84 °C
EINECS:
255-868-5
:
GHS07
Storage Temperature:
Ambient