
6-Chloroimidazo[2,1-b]thiazole-5-sulphonyl chloride
Catalog # APOSOR29800-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:257.103 g/mol
Melting Pt:138…140 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:150020-64-7
Formula:C₅H₂Cl₂N₂O₂S₂
MDL Number:MFCD00205139