Order Entry
Export
ContactUsLinkComponent
N-(3,5-Dichlorophenyl)-2-chloroacetamide
Irritant
N-(3,5-Dichlorophenyl)-2-chloroacetamide
Catalog # APOSOR29761-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(3,5-Dichlorophenyl)-2-chloroacetamide
Catalog # APOSOR29761-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:140…142 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:33560-48-4
Formula:C₈H₆Cl₃NO