Order Entry
Export
Orders LinkContactUsLinkComponent
2-Phenylpropionic acid 97%

2-Phenylpropionic acid 97%

Irritant
Supplier:   Apollo Scientific
2-Phenylpropionic acid 97%
Supplier:  Apollo Scientific
Certificates
2-Phenylpropionic acid 97%

3 Options Available Below

Product Details & Documents
Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:260…266 °C
  • Melting Pt:–5 °C
  • Density:1.1 g/cm³ (25 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:207-752-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,20 €
    36,20 €
    1 * 25 g
       36,20 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 94,10 €
    94,10 €
    1 * 100 g
       94,10 €