Order Entry
Export
ContactUsLinkComponent
4-(Chloromethyl)-2-methyl-1,3-thiazole 95%
Irritant
4-(Chloromethyl)-2-methyl-1,3-thiazole 95%
Catalog # APOSOR29650-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Chloromethyl)-2-methyl-1,3-thiazole 95%
Catalog # APOSOR29650-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.63 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:39238-07-8
 :MFCD00067725
Formula:C₅H₆ClNS
MDL Number:MFCD00051642