Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3,5-Dichlorophenoxy)acetonitrile
(3,5-Dichlorophenoxy)acetonitrile
(3,5-Dichlorophenoxy)acetonitrile
(3,5-Dichlorophenoxy)acetonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
(3,5-Dichlorophenoxy)acetonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
103140-12-1
:
Formula:
C₈H₅Cl₂NO
MDL Number:
MFCD00052707
:
202.04 g/mol
Storage Temperature:
Ambient