Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4'-Dichloroacetanilide
2,4'-Dichloroacetanilide
2,4'-Dichloroacetanilide
APOSOR29570-1G
:
Apollo Scientific
:
2,4'-Dichloroacetanilide
APOSOR29570-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
204.0533 g/mol
:
169…172 °C
:
GHS07
:
Ambient
:
3289-75-6
:
C₈H₇Cl₂NO
:
MFCD00018904