Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,3,5-Trichlorotoluene
α,3,5-Trichlorotoluene 98+%
α,3,5-Trichlorotoluene 98+%
Corrosive
α,3,5-Trichlorotoluene 98+%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
α,3,5-Trichlorotoluene 98+%
Supplier:
Apollo Scientific
About this item
CAS Number:
3290-06-0
:
Formula:
C₇H₅Cl₃
MDL Number:
MFCD00060278
:
195.47 g/mol
Melting Pt:
35…37 °C
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05