Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4,6-dimethoxyaniline
2-Chloro-4,6-dimethoxyaniline 95%
2-Chloro-4,6-dimethoxyaniline 95%
:
Apollo Scientific
2-Chloro-4,6-dimethoxyaniline 95%
:
Apollo Scientific
:
82485-84-5
:
:
C₈H₁₀ClNO₂
:
MFCD01567107
:
187.63 g/mol
:
44…45 °C
:
GHS07
:
Refrigerator