Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thieno[3,2-b]thiophene
Thieno[3,2-b]thiophene 98%
Thieno[3,2-b]thiophene 98%
:
Apollo Scientific
Thieno[3,2-b]thiophene 98%
:
Apollo Scientific
:
251-41-2
:
:
C₆H₄S₂
:
MFCD00179479
:
140.23 g/mol
:
71 °C/1mmHg
:
53…58 °C
:
99 °C
:
Refrigerator
:
3335
:
9,III
:
GHS07