Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,5-dibromobenzoate
Methyl-3,5-dibromobenzoate 98%
Methyl-3,5-dibromobenzoate 98%
Irritant
Methyl-3,5-dibromobenzoate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-3,5-dibromobenzoate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
51329-15-8
:
Formula:
C₈H₆Br₂O₂
MDL Number:
MFCD00082648
:
293.94 g/mol
Melting Pt:
62…63 °C
:
GHS07
Storage Temperature:
Ambient