Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2':5',2''-Terthiophene-5-carboxaldehyde
2,2':5',2''-Terthiophene-5-carboxaldehyde
2,2':5',2''-Terthiophene-5-carboxaldehyde
2,2':5',2''-Terthiophene-5-carboxaldehyde
APOSOR29237-5G
:
Apollo Scientific
:
2,2':5',2''-Terthiophene-5-carboxaldehyde
APOSOR29237-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
276.39 g/mol
:
Ambient
:
7342-41-8
:
C₁₃H₈OS₃
:
MFCD00115181