2-Formylbenzene-1,4-disulphonate
Catalog # APOSOR2923-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.25 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:51818-11-2
Formula:C₇H₆O₇S₂
MDL Number:MFCD03840860