
N-(1,3-Benzodioxol-5-yl)-2-chloroacetamide 97%
Catalog # APOSOR2919-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:213.62 g/mol
Melting Pt:155…156 °C
:GHS07
Storage Temperature:Ambient
CAS Number:227199-07-7
Formula:C₉H₈ClNO₃
MDL Number:MFCD00245737