Tri(2-thienyl)phosphine
Catalog # APOSOR29178-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.36 g/mol
Melting Pt:28…30 °C
:GHS07
Storage Temperature:Ambient
CAS Number:24171-89-9
Formula:C₁₂H₉PS₃
MDL Number:MFCD00015471