

2,5-Dimethylfuran 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:625-86-5
:
Formula:C₆H₈O
MDL Number:MFCD00003250
:96.13 g/mol
Boiling Pt:91…93 °C
Melting Pt:–62 °C
Density:0.903 g/cm³ (25 °C)
Flash Pt:1 °C
Storage Temperature:Ambient
EINECS:210-914-3
UN:1993
ADR:3,II
:GHS02
:GHS07