Isobutyl isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:591-82-2
:
Formula:C₅H₉NS
MDL Number:MFCD00022062
:115.2 g/mol
Boiling Pt:160…162 °C
Density:0.94 g/cm³ (25 °C)
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:209-733-2
UN:3286
ADR:3+6.1+8,II
:GHS05
:GHS07