Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Adamantanol
2-Adamantanol
2-Adamantanol
Irritant
2-Adamantanol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Adamantanol
Supplier:
Apollo Scientific
About this item
CAS Number:
700-57-2
:
Formula:
C₁₀H₁₆O
MDL Number:
MFCD00074744
:
152.24 g/mol
Melting Pt:
258…262 °C
EINECS:
211-846-7
:
GHS07
Storage Temperature:
Ambient