3-Methylpyrocatechol 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol
- CAS Number:488-17-5
- Formula:C₇H₈O₂
- MDL Number:MFCD00016435
- Molecular Weight:124.14 g/mol
- Boiling Pt:241 °C
- Melting Pt:65…68 °C
- Storage Temperature:Ambient
- EINECS:207-672-6
3 Options Available Below
Product Details & Documents
Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol
- CAS Number:488-17-5
- Formula:C₇H₈O₂
- MDL Number:MFCD00016435
- Molecular Weight:124.14 g/mol
- Boiling Pt:241 °C
- Melting Pt:65…68 °C
- Storage Temperature:Ambient
- EINECS:207-672-6



