
3-Hydroxy-p-anisaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:621-59-0
:
Formula:C₈H₈O₃
MDL Number:MFCD00003369
:152.15 g/mol
Boiling Pt:179 °C/15mmHg
Melting Pt:112…116 °C
Density:1.2 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:210-694-9
:GHS07
Storage Temperature:Ambient