Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Cyanopyrazine

Bottle, 5 g
Catalog # APOSOR28727-5G
Irritant
Supplier:   Apollo Scientific
2-Cyanopyrazine
Catalog # APOSOR28727-5G
Supplier:  Apollo Scientific
CAS Number:  19847-12-2
2-Cyanopyrazine

Bottle, 5 g

CAS Number:  19847-12-2
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
  • CAS Number:19847-12-2
  • Formula:C₅H₃N₃
  • MDL Number:MFCD00049361
  • Molecular Weight:105.10 g/mol
  • Boiling Pt:90…93 °C/15mmHg
  • Melting Pt:18…20 °C
  • Density:1.174 g/cm³ (25 °C)
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
  • EINECS:243-369-5
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications