5-Chloro-2-nitroaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1635-61-6
:
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD00007776
:172.57 g/mol
Melting Pt:124…129 °C (dec.)
Storage Temperature:Ambient
EINECS:216-661-5
UN:2237
ADR:6.1,III
:GHS06
:GHS09