
Sesamol-(3,4-(methylenedioxy)phenol)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:533-31-3
:
Formula:C₇H₆O₃
MDL Number:MFCD00005827
:138.12 g/mol
Boiling Pt:113…116 °C/2mmHg
Melting Pt:61…65 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:208-561-5
:GHS07