Order Entry
Export
ContactUsLinkComponent
1,8-Dibromooctane 98%
1,8-Dibromooctane 98%
Catalog # APOSOR28631-100G
Supplier:  Apollo Scientific
CAS Number:  
1,8-Dibromooctane 98%
Catalog # APOSOR28631-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :272.02 g/mol
Boiling Pt:270…272 °C
Melting Pt:15…16 °C
Storage Temperature:Ambient
CAS Number:4549-32-0
Formula:BrH₂C(CH₂)₆CH₂Br
MDL Number:MFCD00000277