
α,2,6-Trichlorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2014-83-7
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00000897
:195.47 g/mol
Boiling Pt:117…119 °C/14mmHg
Melting Pt:36…39 °C
Flash Pt:>113 °C
Storage Temperature:Ambient
EINECS:217-940-4
UN:3261
ADR:8,II
:GHS05