

3,3-Dimethylbutanone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75-97-8
:
Formula:C₆H₁₂O
MDL Number:MFCD00008846
:100.16 g/mol
Boiling Pt:105…106 °C
Melting Pt:–53 °C
Density:0.801 g/cm³ (25 °C)
Flash Pt:5 °C
Storage Temperature:Ambient
UN:1224
ADR:3,II
:GHS02
:GHS07