Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Disodium-2,7-naphthalenedisulphonate
Disodium-2,7-naphthalenedisulphonate
Disodium-2,7-naphthalenedisulphonate
Irritant
Disodium-2,7-naphthalenedisulphonate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Disodium-2,7-naphthalenedisulphonate
Supplier:
Apollo Scientific
About this item
CAS Number:
1655-35-2
:
Formula:
C₁₀H₈O₆S₂
MDL Number:
MFCD00066365
:
332.25 g/mol
EINECS:
216-733-6
:
GHS07
Storage Temperature:
Ambient