Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Disodium-1,6-naphthalenedisulphonate
Disodium-1,6-naphthalenedisulphonate 97%
Disodium-1,6-naphthalenedisulphonate 97%
Irritant
Disodium-1,6-naphthalenedisulphonate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Disodium-1,6-naphthalenedisulphonate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1655-43-2
:
Formula:
C₁₀H₈O₆S₂
MDL Number:
MFCD00021522
:
332.25 g/mol
EINECS:
216-734-1
:
GHS07
Storage Temperature:
Ambient