Order Entry
Export
ContactUsLinkComponent
4-Amino-2,3,5-trichloropyridine 95+%
Irritant
Environment
4-Amino-2,3,5-trichloropyridine 95+%
Catalog # APOSOR27920-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
4-Amino-2,3,5-trichloropyridine 95+%
Catalog # APOSOR27920-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :197.45 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:28443-69-8
Formula:C₅H₃Cl₃N₂
MDL Number:MFCD00153908