Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-hydroxycyclohexanecarboxylate
Methyl-4-hydroxycyclohexanecarboxylate 95%
Methyl-4-hydroxycyclohexanecarboxylate 95%
:
Apollo Scientific
Methyl-4-hydroxycyclohexanecarboxylate 95%
:
Apollo Scientific
:
17449-76-2
:
:
C₈H₁₄O₃
:
MFCD00729087
:
158.2 g/mol
:
GHS07
:
Ambient