Order Entry
Export
ContactUsLinkComponent
Methyl-2,6-dihydroxybenzoate 98%
Irritant
Methyl-2,6-dihydroxybenzoate 98%
Catalog # APOSOR27592-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2,6-dihydroxybenzoate 98%
Catalog # APOSOR27592-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :168.15 g/mol
Melting Pt:67…70 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:2150-45-0
Formula:C₈H₈O₄
MDL Number:MFCD00059619