Order Entry
Export
ContactUsLinkComponent
2,6-Dimethyl-4-pyrone
Irritant
2,6-Dimethyl-4-pyrone
Catalog # APOSOR27582-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dimethyl-4-pyrone
Catalog # APOSOR27582-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:248…250 °C
Melting Pt:132…137 °C
 :124.14 g/mol
EINECS:213-719-1
 :GHS07
Storage Temperature:Ambient
CAS Number:1004-36-0
Formula:C₇H₈O₂
MDL Number:MFCD00006579