
6-Bromo-8-nitroimidazo[1,2-a]pyridine
Catalog # APOSOR2749-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.04 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:1031927-16-8
Formula:C₇H₄BrN₃O₂
MDL Number:MFCD09836167