Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromo-4-hydroxybenzaldehyde
3,5-Dibromo-4-hydroxybenzaldehyde 98%
3,5-Dibromo-4-hydroxybenzaldehyde 98%
Irritant
3,5-Dibromo-4-hydroxybenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dibromo-4-hydroxybenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
2973-77-5
:
Formula:
C₇H₄Br₂O₂
MDL Number:
MFCD00016980
:
279.92 g/mol
Melting Pt:
181…185 °C
:
GHS07
Storage Temperature:
Ambient