Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dibromopyridazine
3,6-Dibromopyridazine
3,6-Dibromopyridazine
Irritant
3,6-Dibromopyridazine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,6-Dibromopyridazine
Supplier:
Apollo Scientific
About this item
CAS Number:
17973-86-3
:
Formula:
C₄H₂Br₂N₂
MDL Number:
MFCD00233947
:
237.88 g/mol
Melting Pt:
117…121 °C
:
GHS07
Storage Temperature:
Ambient