Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 5-bromonicotinate
Methyl-5-bromonicotinate
Methyl-5-bromonicotinate
:
Apollo Scientific
Methyl-5-bromonicotinate
:
Apollo Scientific
:
29681-44-5
:
:
C₇H₆BrNO₂
:
MFCD00173668
:
216.03 g/mol
:
98…100 °C
:
GHS07
:
Ambient