Methyl 5,6-dichloronicotinate 97%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Methyl 5,6-dichloronicotinate
- CAS Number:56055-54-0
- Formula:C₇H₅Cl₂NO₂
- MDL Number:MFCD00125367
- Molecular Weight:206.03 g/mol
- Melting Pt:68…70 °C
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
Synonyms: Methyl 5,6-dichloronicotinate
- CAS Number:56055-54-0
- Formula:C₇H₅Cl₂NO₂
- MDL Number:MFCD00125367
- Molecular Weight:206.03 g/mol
- Melting Pt:68…70 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



