Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Diiodothiophene 99%

2,5-Diiodothiophene 99%

Irritant
Supplier:   Apollo Scientific
2,5-Diiodothiophene 99%
Supplier:  Apollo Scientific
Certificates
2,5-Diiodothiophene 99%

5 Options Available Below

Product Details & Documents
  • CAS Number:625-88-7
  • Formula:C₄H₂I₂S
  • MDL Number:MFCD00014525
  • Molecular Weight:335.93 g/mol
  • Boiling Pt:139…140 °C
  • Melting Pt:41…42 °C
  • Flash Pt:96 °C
  • Storage Temperature:Refrigerator
  • EINECS:210-915-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 37,70 €
    37,70 €
    1 * 5 g
       37,70 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 186,00 €
    186,00 €
    1 * 25 g
       186,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 738,00 €
    738,00 €
    1 * 100 g
       738,00 €

    Specifications:
    New Product
    New Product