Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(3,5-Dichlorophenyl)propene
2-(3,5-Dichlorophenyl)propene 99%
2-(3,5-Dichlorophenyl)propene 99%
2-(3,5-Dichlorophenyl)propene 99%
APOSOR2733-1G
:
Apollo Scientific
:
2-(3,5-Dichlorophenyl)propene 99%
APOSOR2733-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
187.07 g/mol
:
GHS07
:
Ambient
:
68575-36-0
:
C₉H₈Cl₂
:
MFCD00035987