Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-1,2-benzisoxazol-3-ol
6-Chloro-1,2-benzisoxazol-3-ol
6-Chloro-1,2-benzisoxazol-3-ol
6-Chloro-1,2-benzisoxazol-3-ol
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloro-1,2-benzisoxazol-3-ol
Supplier:
Apollo Scientific
About this item
CAS Number:
61977-29-5
:
Formula:
C₇H₄ClNO₂
MDL Number:
MFCD00125033
:
169.57 g/mol
Melting Pt:
215…217 °C
:
GHS07
Storage Temperature:
Ambient