Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dichloroisonicotinamide

Bottle, 250 mg
Catalog # APOSOR27131-250MG
Irritant
Supplier:   Apollo Scientific
2,6-Dichloroisonicotinamide
Catalog # APOSOR27131-250MG
Supplier:  Apollo Scientific
CAS Number:  89281-13-0
2,6-Dichloroisonicotinamide

Bottle, 250 mg

CAS Number:  89281-13-0
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:89281-13-0
  • Formula:C₆H₄Cl₂N₂O
  • MDL Number:MFCD00052640
  • Molecular Weight:191.0200 g/mol
  • Melting Pt:202…206 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications