
2,6-Dichloroisonicotinoyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42521-08-4
:
Formula:C₆H₂Cl₃NO
MDL Number:MFCD00052639
:210.45 g/mol
Boiling Pt:140 °C/15mmHg
Melting Pt:25 °C
Density:1.537 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05