
4,6-Dimethylpyridin-2-ol
Catalog # APOSOR27029-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:178…179 °C
:123.16 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:16115-08-5
Formula:C₇H₉NO
MDL Number:MFCD00831528