Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2,3-Dichlorophenyl)-2-chloroacetamide
N-(2,3-Dichlorophenyl)-2-chloroacetamide
N-(2,3-Dichlorophenyl)-2-chloroacetamide
:
Apollo Scientific
N-(2,3-Dichlorophenyl)-2-chloroacetamide
:
Apollo Scientific
:
33560-47-3
:
:
C₈H₆Cl₃NO
:
104…106 °C
:
GHS07
:
Ambient