
3,4-Dibutoxy-3-cyclobutene-1,2-dione 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2892-62-8
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00037150
:226.27 g/mol
Boiling Pt:155…158 °C
Density:1.047 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient