
4-(4-Methoxyphenyl)-1,3-thiazol-2-amine
Catalog # APOSOR26726-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:206.27 g/mol
Melting Pt:206…209 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:2104-04-3
Formula:C₁₀H₁₀N₂OS
MDL Number:MFCD00170663