1,3-Adamantanediol
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,3-Dihydroxyadamantane
- CAS Number:5001-18-3
- Formula:C₁₀H₁₆O₂
- MDL Number:MFCD00154013
- Molecular Weight:168.24 g/mol
- Melting Pt:260 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 1,3-Dihydroxyadamantane
- CAS Number:5001-18-3
- Formula:C₁₀H₁₆O₂
- MDL Number:MFCD00154013
- Molecular Weight:168.24 g/mol
- Melting Pt:260 °C
- Storage Temperature:Ambient



