
1,2-Diiodoethane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:624-73-7
:
Formula:C₂H₄I₂
MDL Number:MFCD00001093
:281.86 g/mol
Melting Pt:80…84 °C
Density:2.132 g/cm³ (25 °C)
EINECS:210-859-5
:GHS07
Storage Temperature:Refrigerator