Order Entry
Export
ContactUsLinkComponent
4-Methoxy-1,2-benzisoxazol-3-amine
Irritant
4-Methoxy-1,2-benzisoxazol-3-amine
Catalog # APOSOR25957-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxy-1,2-benzisoxazol-3-amine
Catalog # APOSOR25957-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :164.160 g/mol
Melting Pt:140…142 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:177995-40-3
Formula:C₈H₈N₂O₂
MDL Number:MFCD03407354